The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000345.3 | NP_000336.1 | 2 - 3 | 384 | 62 |
NM_001146054.1 | NP_001139526.1 | 2 - 3 | 380 | 62 | |
NM_001146055.1 | NP_001139527.1 | 2 - 3 | 191 | 62 | |
NM_007308.2 | NP_009292.1 | 2 - 3 | 380 | 62 | |
XM_011532203.1 | XP_011530505.1 | 2 - 3 | 413 | 62 | |
XM_011532204.2 | XP_011530506.1 | 2 - 3 | 517 | 62 | |
XM_011532205.2 | XP_011530507.1 | 2 - 3 | 219 | 62 | |
XM_011532206.1 | XP_011530508.1 | 2 - 3 | 241 | 62 | |
XM_011532207.1 | XP_011530509.1 | 2 - 3 | 228 | 62 | |
XM_011532208.2 | XP_011530510.1 | 2 - 3 | 261 | 62 | |
XM_017008562.1 | XP_016864051.1 | 2 - 3 | 255 | 62 | |
XM_017008563.1 | XP_016864052.1 | 2 - 3 | 281 | 62 | |
GenBank mRNA | AK290169.1 | - | 2 - 3 | 205 | 62 |
AY049786.1 | - | 1 - 2 | 123 | 62 | |
BC013293.2 | - | 2 - 3 | 544 | 62 | |
BC108275.1 | - | 2 - 3 | 338 | 62 | |
CR457058.1 | - | 1 - 2 | 121 | 62 | |
CR541653.1 | - | 1 - 2 | 121 | 62 | |
CR736451.1 | - | 2 - 3 | 380 | 62 | |
D31839.1 | - | 1 - 2 | 134 | 62 | |
DA391115.1 | - | 2 - 3 | 191 | 62 | |
HQ830267.1 | - | 2 - 3 | 393 | 62 | |
HQ830268.1 | - | 2 - 3 | 556 | 62 | |
HQ830269.1 | - | 2 - 3 | 246 | 62 | |
JN709861.1 | - | 2 - 3 | 384 | 62 | |
JN709864.1 | - | 2 - 3 | 191 | 62 | |
JN709867.1 | - | 2 - 3 | 233 | 62 | |
L08850.1 | - | 2 - 3 | 173 | 62 | |
L36675.1 | - | 1 - 2 | 139 | 62 |