The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000345.3 | NP_000336.1 | 5 - 6 | 654 | 97 |
NM_001146054.1 | NP_001139526.1 | 5 - 6 | 650 | 97 | |
NM_001146055.1 | NP_001139527.1 | 5 - 6 | 461 | 97 | |
XM_011532208.2 | XP_011530510.1 | 5 - 6 | 531 | 97 | |
XM_017008562.1 | XP_016864051.1 | 5 - 6 | 525 | 97 | |
XM_017008563.1 | XP_016864052.1 | 5 - 6 | 551 | 97 | |
GenBank mRNA | AK290169.1 | - | 5 - 6 | 475 | 97 |
BC013293.2 | - | 5 - 6 | 814 | 97 | |
BC108275.1 | - | 5 - 6 | 608 | 97 | |
DA391115.1 | - | 5 - 6 | 461 | 97 | |
JN709859.1 | - | 4 - 5 | 608 | 97 | |
JN709862.1 | - | 4 - 5 | 612 | 97 | |
JN709865.1 | - | 4 - 5 | 419 | 97 | |
JN709868.1 | - | 4 - 5 | 461 | 97 | |
L08850.1 | - | 5 - 6 | 443 | 97 |