The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020236.3 | NP_064621.3 | 8 - 9 | 1060 | 99 |
XM_011532198.2 | XP_011530500.1 | 9 - 10 | 1112 | 99 | |
XM_011532199.1 | XP_011530501.1 | 9 - 10 | 926 | 99 | |
GenBank mRNA | AB049474.1 | - | 7 - 8 | 834 | 99 |
AF212225.1 | - | 8 - 9 | 964 | 99 | |
AK024791.1 | - | 6 - 7 | 663 | 99 | |
AK315600.1 | - | 8 - 9 | 877 | 99 | |
BC014356.1 | - | 8 - 9 | 884 | 99 | |
BC015109.1 | - | 8 - 9 | 873 | 99 | |
BC017765.1 | - | 8 - 9 | 881 | 99 | |
BC032595.1 | - | 8 - 9 | 871 | 99 | |
BU675164.1 | - | 1 - 2 | 99 | ||
CR457191.1 | - | 7 - 8 | 796 | 99 |