The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000716.3 | NP_000707.1 | 1 - 2 | 353 | 65 |
NM_001017365.2 | NP_001017365.1 | 2 - 3 | 215 | 65 | |
NM_001017367.1 | NP_001017367.1 | 2 - 3 | 189 | 65 | |
XM_005273254.4 | XP_005273311.1 | 3 - 4 | 494 | 65 | |
GenBank mRNA | H63461.1 | - | 2 - 3 | 189 | 65 |
L11245.1 | - | 1 - 2 | 338 | 65 | |
M29964.1 | - | 1 - 2 | 88 | 65 |