Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL13A1
Entrez Gene ID
1305
Gene Name
collagen type XIII alpha 1 chain
Gene Aliases
CMS19, COLXIIIA1
Chromosome Location
Chr.10: 69801885 - 69959148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130103.1 NP_001123575.1 35 - 36 2473 65
NM_001320951.1 NP_001307880.1 33 - 34 2362 65
NM_080798.3 NP_542988.3 30 - 31 2257 65
NM_080800.3 NP_542990.3 33 - 34 2380 65
NM_080801.3 NP_542991.3 34 - 35 2407 65
NM_080802.3 NP_542992.3 32 - 33 2326 65
NM_080805.3 NP_542995.3 30 - 31 2239 65
XM_011539292.2 XP_011537594.1 36 - 37 2509 65
XM_011539293.2 XP_011537595.1 35 - 36 2464 65
XM_011539294.2 XP_011537596.1 35 - 36 2446 65
XM_011539295.2 XP_011537597.1 34 - 35 2428 65
XM_017015676.1 XP_016871165.1 36 - 37 2509 65
XM_017015677.1 XP_016871166.1 35 - 36 2473 65
XM_017015678.1 XP_016871167.1 34 - 35 2419 65
XM_017015679.1 XP_016871168.1 34 - 35 2428 65
XM_017015680.1 XP_016871169.1 34 - 35 2422 65
XM_017015681.1 XP_016871170.1 34 - 35 2401 65
XM_017015682.1 XP_016871171.1 33 - 34 2386 65
XM_017015683.1 XP_016871172.1 33 - 34 2386 65
XM_017015684.1 XP_016871173.1 33 - 34 2365 65
XM_017015685.1 XP_016871174.1 33 - 34 2359 65
XM_017015686.1 XP_016871175.1 33 - 34 2359 65
XM_017015687.1 XP_016871176.1 34 - 35 2428 65
XM_017015688.1 XP_016871177.1 32 - 33 2323 65
XM_017015689.1 XP_016871178.1 34 - 35 2401 65
XM_017015690.1 XP_016871179.1 30 - 31 2296 65
XM_017015691.1 XP_016871180.1 33 - 34 2365 65
XM_017015692.1 XP_016871181.1 31 - 32 2287 65
XM_017015693.1 XP_016871182.1 33 - 34 2359 65
XM_017015694.1 XP_016871183.1 29 - 30 2218 65
XM_017015695.1 XP_016871184.1 29 - 30 2218 65
XM_017015696.1 XP_016871185.1 32 - 33 1599 65
XM_017015697.1 XP_016871186.1 32 - 33 1796 65
GenBank mRNA AJ293624.1 - 35 - 36 2434 65
BC136385.1 - 30 - 31 2027 65
BC139913.1 - 6 - 7 1025 65
BY796799.2 - 5 - 6 274 65
M15524.1 - 4 - 5 186 65
M33653.1 - 9 - 10 523 65
M59217.1 - 34 - 35 1740 65

View Full Product Details