Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL13A1
Entrez Gene ID
1305
Gene Name
collagen type XIII alpha 1 chain
Gene Aliases
CMS19, COLXIIIA1
Chromosome Location
Chr.10: 69801885 - 69959148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130103.1 NP_001123575.1 6 - 7 1022 71
NM_001320951.1 NP_001307880.1 7 - 8 1049 71
NM_080800.3 NP_542990.3 6 - 7 1022 71
NM_080801.3 NP_542991.3 6 - 7 1022 71
NM_080802.3 NP_542992.3 6 - 7 1022 71
XM_011539292.2 XP_011537594.1 7 - 8 1052 71
XM_011539293.2 XP_011537595.1 7 - 8 1052 71
XM_011539294.2 XP_011537596.1 7 - 8 1052 71
XM_011539295.2 XP_011537597.1 7 - 8 1052 71
XM_017015676.1 XP_016871165.1 7 - 8 1052 71
XM_017015677.1 XP_016871166.1 7 - 8 1052 71
XM_017015678.1 XP_016871167.1 6 - 7 1025 71
XM_017015679.1 XP_016871168.1 7 - 8 1052 71
XM_017015680.1 XP_016871169.1 7 - 8 1052 71
XM_017015681.1 XP_016871170.1 7 - 8 1052 71
XM_017015682.1 XP_016871171.1 7 - 8 1052 71
XM_017015683.1 XP_016871172.1 6 - 7 1016 71
XM_017015684.1 XP_016871173.1 6 - 7 1025 71
XM_017015685.1 XP_016871174.1 7 - 8 1052 71
XM_017015686.1 XP_016871175.1 7 - 8 1052 71
XM_017015687.1 XP_016871176.1 7 - 8 1052 71
XM_017015688.1 XP_016871177.1 7 - 8 1052 71
XM_017015689.1 XP_016871178.1 7 - 8 1052 71
XM_017015690.1 XP_016871179.1 4 - 5 962 71
XM_017015691.1 XP_016871180.1 7 - 8 1052 71
XM_017015692.1 XP_016871181.1 6 - 7 1016 71
XM_017015693.1 XP_016871182.1 7 - 8 1052 71
XM_017015696.1 XP_016871185.1 4 - 5 205 71
XM_017015697.1 XP_016871186.1 6 - 7 483 71
GenBank mRNA AJ293624.1 - 6 - 7 71
M59217.1 - 6 - 7 71

View Full Product Details