Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPC
Entrez Gene ID
7508
Gene Name
XPC complex subunit, DNA damage recognition and repair factor
Gene Aliases
RAD4, XP3, XPCC, p125
Chromosome Location
Chr.3: 14145147 - 14178672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004628.4 NP_004619.3 11 - 12 2220 65
NR_027299.1 10 - 11 2096 65
GenBank mRNA AK222844.1 - 10 - 11 1913 65
AK289761.1 - 11 - 12 2149 65
AK295711.1 - 11 - 12 2038 65
AK311039.1 - 10 - 11 2026 65
BC016620.1 - 11 - 12 2131 65
D21089.1 - 11 - 12 2221 65
EU530524.1 - 2 - 3 400 65
EU530526.1 - 2 - 3 400 65
EU530529.1 - 3 - 4 427 65
EU530532.1 - 3 - 4 576 65
EU530534.1 - 3 - 4 427 65
HM113488.1 - 11 - 12 2120 65
KJ534962.1 - 11 - 12 2116 65
KJ535085.1 - 9 - 10 1832 65
X65024.1 - 10 - 11 1956 65

View Full Product Details