The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128159.2 | NP_001121631.1 | 19 - 20 | 2235 | 77 |
XM_017024817.1 | XP_016880306.1 | 16 - 17 | 2281 | 77 | |
XM_017024818.1 | XP_016880307.1 | 15 - 16 | 2476 | 77 | |
GenBank mRNA | AK091388.1 | - | 14 - 15 | 1858 | 77 |
EU021218.1 | - | 19 - 20 | 2088 | 77 |