The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291958.1 | NP_001278887.1 | 11 - 12 | 3123 | 74 |
NM_001301072.1 | NP_001288001.1 | 10 - 11 | 3025 | 74 | |
NM_005922.3 | NP_005913.2 | 10 - 11 | 3025 | 74 | |
NM_006724.3 | NP_006715.2 | 10 - 11 | 3025 | 74 | |
NR_120425.1 | 10 - 11 | 3025 | 74 | ||
XM_005266989.2 | XP_005267046.1 | 10 - 11 | 3229 | 74 | |
XM_017010869.1 | XP_016866358.1 | 11 - 12 | 3449 | 74 | |
GenBank mRNA | AF002715.1 | - | 10 - 11 | 2962 | 74 |
AK307866.1 | - | 10 - 11 | 2894 | 74 | |
BC136276.1 | - | 10 - 11 | 2912 | 74 | |
BC143735.1 | - | 10 - 11 | 2909 | 74 | |
BC143743.1 | - | 10 - 11 | 2908 | 74 | |
BC143750.1 | - | 11 - 12 | 3006 | 74 | |
BC146770.1 | - | 10 - 11 | 3025 | 74 | |
D86968.2 | - | 10 - 11 | 3025 | 74 |