The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011544026.2 | XP_011542328.1 | 14 - 15 | 1769 | 127 |
XM_011544028.2 | XP_011542330.1 | 12 - 13 | 1544 | 127 | |
XM_017000105.1 | XP_016855594.1 | 13 - 14 | 1774 | 127 | |
GenBank mRNA | AF039690.1 | - | 7 - 8 | 958 | 127 |