Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 7 - 8 911 137
XM_005273013.4 XP_005273070.1 6 - 7 743 137
XM_005273018.1 XP_005273075.1 6 - 7 750 137
XM_005273023.4 XP_005273080.1 7 - 8 1588 137
XM_006711727.3 XP_006711790.1 7 - 8 797 137
XM_006711728.3 XP_006711791.1 6 - 7 668 137
XM_011544021.2 XP_011542323.1 8 - 9 893 137
XM_011544022.2 XP_011542324.1 8 - 9 968 137
XM_011544023.2 XP_011542325.1 8 - 9 893 137
XM_011544024.2 XP_011542326.1 8 - 9 893 137
XM_011544026.2 XP_011542328.1 8 - 9 893 137
XM_011544028.2 XP_011542330.1 6 - 7 668 137
XM_011544029.2 XP_011542331.1 8 - 9 893 137
XM_017000103.1 XP_016855592.1 8 - 9 893 137
XM_017000104.1 XP_016855593.1 6 - 7 743 137
XM_017000105.1 XP_016855594.1 7 - 8 898 137
XM_017000106.1 XP_016855595.1 7 - 8 879 137
XM_017000109.1 XP_016855598.1 7 - 8 797 137
GenBank mRNA AF039690.1 - 1 - 2 82 137
AF161348.1 - 7 - 8 888 137
AF250731.1 - 7 - 8 742 137
AK297105.1 - 3 - 4 330 137
AK299249.1 - 4 - 5 503 137
BC032454.1 - 7 - 8 854 137

View Full Product Details