The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 10 - 11 | 1052 | 125 |
NM_003618.3 | NP_003609.2 | 10 - 11 | 1052 | 125 | |
GenBank mRNA | AK314487.1 | - | 10 - 11 | 889 | 125 |
BC071579.1 | - | 10 - 11 | 1044 | 125 |