The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 20 - 21 | 1845 | 86 |
NM_003618.3 | NP_003609.2 | 21 - 22 | 1908 | 86 | |
GenBank mRNA | AK291467.1 | - | 21 - 22 | 1874 | 86 |
AK299889.1 | - | 13 - 14 | 937 | 86 | |
AK314487.1 | - | 20 - 21 | 1682 | 86 | |
BC071579.1 | - | 20 - 21 | 1837 | 86 |