The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004789.3 | NP_004780.3 | 2 - 3 | 1065 | 112 |
XM_006717323.2 | XP_006717386.1 | 2 - 3 | 925 | 112 | |
GenBank mRNA | BC034458.1 | - | 2 - 3 | 839 | 112 |
BC093662.1 | - | 2 - 3 | 405 | 112 | |
BC112185.1 | - | 2 - 3 | 405 | 112 |