The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284413.1 | NP_001271342.1 | 12 - 13 | 1555 | 63 |
NM_001284415.1 | NP_001271344.1 | 11 - 12 | 1456 | 63 | |
NM_001284416.1 | NP_001271345.1 | 10 - 11 | 1390 | 63 | |
NM_012260.3 | NP_036392.2 | 13 - 14 | 1636 | 63 | |
NR_104315.1 | 11 - 12 | 1457 | 63 | ||
GenBank mRNA | AF161397.1 | - | 13 - 14 | 1256 | 63 |
AJ131753.1 | - | 13 - 14 | 1346 | 63 | |
AK056977.1 | - | 5 - 6 | 630 | 63 | |
AK301546.1 | - | 12 - 13 | 1272 | 63 | |
AK301990.1 | - | 10 - 11 | 1125 | 63 | |
AK302086.1 | - | 11 - 12 | 1173 | 63 | |
BC001627.1 | - | 13 - 14 | 1354 | 63 | |
BC007440.2 | - | 11 - 12 | 1162 | 63 |