The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008541.1 | NP_001008541.1 | 3 - 4 | 326 | 131 |
NM_005962.4 | NP_005953.4 | 4 - 5 | 551 | 131 | |
NM_130439.3 | NP_569157.2 | 4 - 5 | 749 | 131 | |
GenBank mRNA | AK303949.1 | - | 4 - 5 | 428 | 131 |
AK307090.1 | - | 5 - 6 | 437 | 131 | |
AK309718.1 | - | 4 - 5 | 364 | 131 | |
AL538051.3 | - | 4 - 5 | 562 | 131 | |
BC012907.1 | - | 3 - 4 | 326 | 131 | |
BC035128.1 | - | 4 - 5 | 755 | 131 | |
BG700036.1 | - | 4 - 5 | 535 | 131 | |
BM927870.1 | - | 4 - 5 | 534 | 131 | |
BT007069.1 | - | 2 - 3 | 206 | 131 | |
BX417051.2 | - | 4 - 5 | 547 | 131 | |
CR536576.1 | - | 4 - 5 | 344 | 131 | |
D63940.1 | - | 3 - 4 | 236 | 131 | |
KU178207.1 | - | 2 - 3 | 206 | 131 | |
KU178208.1 | - | 3 - 4 | 252 | 131 | |
KU178209.1 | - | 3 - 4 | 236 | 131 |