The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001851.2 | NP_001001851.1 | 10 - 11 | 2083 | 104 |
NM_030569.6 | NP_085046.5 | 10 - 11 | 2083 | 104 | |
NM_032817.5 | NP_116206.4 | 6 - 7 | 1418 | 104 | |
XM_011519713.2 | XP_011518015.1 | 11 - 12 | 2158 | 104 | |
XM_011519714.2 | XP_011518016.1 | 11 - 12 | 2158 | 104 | |
GenBank mRNA | AB075833.1 | - | 7 - 8 | 1624 | 104 |
AF318347.1 | - | 2 - 3 | 968 | 104 | |
AK027375.1 | - | 10 - 11 | 2020 | 104 | |
AK027831.1 | - | 7 - 8 | 1602 | 104 | |
AK027849.1 | - | 5 - 6 | 1267 | 104 | |
AK075381.1 | - | 10 - 11 | 2056 | 104 | |
AL833203.1 | - | 10 - 11 | 1928 | 104 | |
AY238437.1 | - | 10 - 11 | 1979 | 104 | |
BC136439.1 | - | 10 - 11 | 2056 | 104 |