The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318336.1 | NP_001305265.1 | 23 - 24 | 3552 | 68 |
NM_001318337.1 | NP_001305266.1 | 23 - 24 | 3792 | 68 | |
NM_001318338.1 | NP_001305267.1 | 13 - 14 | 2586 | 68 | |
NM_018027.4 | NP_060497.3 | 24 - 25 | 3492 | 68 | |
XM_006717458.3 | XP_006717521.1 | 25 - 26 | 3873 | 68 | |
XM_017016393.1 | XP_016871882.1 | 24 - 25 | 3837 | 68 | |
GenBank mRNA | AB037715.1 | - | 24 - 25 | 3492 | 68 |
BC151244.1 | - | 24 - 25 | 3503 | 68 |