The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290268.1 | NP_001277197.1 | 7 - 8 | 977 | 101 |
NM_080829.3 | NP_543019.2 | 7 - 8 | 910 | 101 | |
NR_110890.1 | 7 - 8 | 1162 | 101 | ||
XM_005260294.3 | XP_005260351.1 | 8 - 9 | 974 | 101 | |
XM_006723713.3 | XP_006723776.1 | 7 - 8 | 1022 | 101 | |
XM_011528578.2 | XP_011526880.1 | 8 - 9 | 1002 | 101 | |
XM_011528579.2 | XP_011526881.1 | 8 - 9 | 1140 | 101 | |
XM_011528580.2 | XP_011526882.1 | 8 - 9 | 1194 | 101 | |
XM_011528581.2 | XP_011526883.1 | 8 - 9 | 1178 | 101 | |
XM_011528584.2 | XP_011526886.1 | 7 - 8 | 1039 | 101 | |
XM_011528586.2 | XP_011526888.1 | 7 - 8 | 1040 | 101 | |
XM_017027682.1 | XP_016883171.1 | 7 - 8 | 1039 | 101 | |
GenBank mRNA | AK056792.1 | - | 7 - 8 | 881 | 101 |
AK295781.1 | - | 7 - 8 | 853 | 101 | |
AK299337.1 | - | 7 - 8 | 826 | 101 |