The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290268.1 | NP_001277197.1 | 18 - 19 | 2778 | 58 |
NM_080829.3 | NP_543019.2 | 18 - 19 | 2711 | 58 | |
NR_110890.1 | 18 - 19 | 2937 | 58 | ||
XM_005260294.3 | XP_005260351.1 | 19 - 20 | 2775 | 58 | |
XM_006723713.3 | XP_006723776.1 | 18 - 19 | 2823 | 58 | |
XM_011528578.2 | XP_011526880.1 | 19 - 20 | 2803 | 58 | |
XM_011528579.2 | XP_011526881.1 | 19 - 20 | 2941 | 58 | |
XM_011528580.2 | XP_011526882.1 | 19 - 20 | 2995 | 58 | |
XM_011528581.2 | XP_011526883.1 | 19 - 20 | 2979 | 58 | |
XM_011528585.2 | XP_011526887.1 | 8 - 9 | 2126 | 58 | |
XM_017027681.1 | XP_016883170.1 | 10 - 11 | 1771 | 58 | |
GenBank mRNA | AK299337.1 | - | 18 - 19 | 2627 | 58 |
AL732057.1 | - | 2 - 3 | 58 | ||
DT932687.1 | - | 5 - 6 | 664 | 58 | |
HY221639.1 | - | 1 - 2 | 58 | ||
HY285972.1 | - | 2 - 3 | 291 | 58 |