The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290268.1 | NP_001277197.1 | 21 - 22 | 3146 | 61 |
NM_080829.3 | NP_543019.2 | 21 - 22 | 3079 | 61 | |
NR_110890.1 | 21 - 22 | 3305 | 61 | ||
XM_005260294.3 | XP_005260351.1 | 22 - 23 | 3143 | 61 | |
XM_006723713.3 | XP_006723776.1 | 21 - 22 | 3191 | 61 | |
XM_011528578.2 | XP_011526880.1 | 22 - 23 | 3171 | 61 | |
XM_011528579.2 | XP_011526881.1 | 22 - 23 | 3309 | 61 | |
XM_011528580.2 | XP_011526882.1 | 22 - 23 | 3363 | 61 | |
XM_011528581.2 | XP_011526883.1 | 22 - 23 | 3347 | 61 | |
XM_011528585.2 | XP_011526887.1 | 11 - 12 | 2494 | 61 | |
XM_017027681.1 | XP_016883170.1 | 13 - 14 | 2139 | 61 | |
GenBank mRNA | AK299337.1 | - | 21 - 22 | 2995 | 61 |
CA389799.1 | - | 2 - 3 | 131 | 61 | |
CD558804.1 | - | 2 - 3 | 173 | 61 |