The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021035.2 | NP_066363.1 | 10 - 11 | 3202 | 96 |
XM_006723836.1 | XP_006723899.1 | 10 - 11 | 3032 | 96 | |
XM_011528924.1 | XP_011527226.1 | 9 - 10 | 3077 | 96 | |
XM_017027971.1 | XP_016883460.1 | 8 - 9 | 2128 | 96 | |
GenBank mRNA | AB037825.2 | - | 10 - 11 | 3020 | 96 |
AK023836.1 | - | 6 - 7 | 954 | 96 | |
AU140771.1 | - | 2 - 3 | 279 | 96 | |
BC041630.1 | - | 3 - 4 | 486 | 96 |