The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134477.2 | NP_001127949.1 | 16 - 17 | 1148 | 78 |
NM_001242992.1 | NP_001229921.1 | 15 - 16 | 1093 | 78 | |
NM_002582.3 | NP_002573.1 | 16 - 17 | 1231 | 78 | |
XM_011522510.2 | XP_011520812.1 | 16 - 17 | 1237 | 78 | |
XM_011522511.2 | XP_011520813.1 | 16 - 17 | 1236 | 78 | |
XM_011522513.2 | XP_011520815.1 | 16 - 17 | 1147 | 78 | |
XM_011522514.1 | XP_011520816.1 | 16 - 17 | 1235 | 78 | |
XM_017023258.1 | XP_016878747.1 | 15 - 16 | 1157 | 78 | |
XM_017023259.1 | XP_016878748.1 | 12 - 13 | 836 | 78 | |
XM_017023260.1 | XP_016878749.1 | 10 - 11 | 725 | 78 | |
GenBank mRNA | AJ005698.1 | - | 16 - 17 | 1142 | 78 |
AK023866.1 | - | 9 - 10 | 516 | 78 | |
AK293189.1 | - | 16 - 17 | 1140 | 78 | |
AK299653.1 | - | 9 - 10 | 673 | 78 | |
AK301648.1 | - | 15 - 16 | 1083 | 78 | |
AK315020.1 | - | 16 - 17 | 1227 | 78 | |
BC044763.1 | - | 16 - 17 | 1169 | 78 | |
BC050029.1 | - | 16 - 17 | 1169 | 78 |