The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194259.2 | NP_919235.1 | 2 - 3 | 403 | 98 |
XM_011522644.2 | XP_011520946.1 | 2 - 3 | 383 | 98 | |
GenBank mRNA | BE262845.1 | - | 2 - 3 | 385 | 98 |
BF315618.1 | - | 2 - 3 | 362 | 98 |