Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP7D2
Entrez Gene ID
256714
Gene Name
MAP7 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 20006713 - 20116996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168465.1 NP_001161937.1 8 - 9 1126 90
NM_001168466.1 NP_001161938.1 6 - 7 868 90
NM_001168467.1 NP_001161939.1 7 - 8 840 90
NM_152780.3 NP_689993.2 7 - 8 1003 90
XM_005274478.2 XP_005274535.1 8 - 9 989 90
XM_005274479.2 XP_005274536.1 8 - 9 956 90
XM_005274480.3 XP_005274537.1 7 - 8 890 90
XM_006724481.2 XP_006724544.1 9 - 10 1136 90
XM_006724482.2 XP_006724545.1 8 - 9 1013 90
XM_011545483.1 XP_011543785.1 9 - 10 1103 90
XM_011545484.1 XP_011543786.1 9 - 10 1086 90
XM_011545485.1 XP_011543787.1 9 - 10 1021 90
XM_011545486.2 XP_011543788.1 9 - 10 1136 90
XM_017029393.1 XP_016884882.1 8 - 9 980 90
XM_017029394.1 XP_016884883.1 8 - 9 963 90
XM_017029395.1 XP_016884884.1 8 - 9 939 90
XM_017029396.1 XP_016884885.1 8 - 9 898 90
XM_017029397.1 XP_016884886.1 8 - 9 874 90
XM_017029398.1 XP_016884887.1 7 - 8 890 90
GenBank mRNA AK027409.1 - 7 - 8 905 90
AK127056.1 - 5 - 6 542 90
AK294788.1 - 7 - 8 834 90
AK296319.1 - 6 - 7 779 90
AK316117.1 - 6 - 7 776 90
BC037165.2 - 5 - 6 670 90
BC089400.1 - 8 - 9 1047 90
BC136379.1 - 7 - 8 920 90
KP849473.1 - 8 - 9 961 90
KP849474.1 - 8 - 9 928 90
KP979782.1 - 8 - 9 995 90
KP979783.1 - 8 - 9 962 90
KP979784.1 - 8 - 9 1019 90
KP979785.1 - 8 - 9 986 90
KP979786.1 - 8 - 9 1019 90

View Full Product Details