The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048205.1 | NP_001041670.1 | 10 - 11 | 1341 | 153 |
NM_005132.2 | NP_005123.2 | 11 - 12 | 1240 | 153 | |
XM_011537421.1 | XP_011535723.1 | 10 - 11 | 1513 | 153 | |
GenBank mRNA | AF006264.1 | - | 10 - 11 | 813 | 153 |
AK001435.1 | - | 11 - 12 | 1240 | 153 | |
AK291191.1 | - | 11 - 12 | 1372 | 153 | |
BC004159.2 | - | 10 - 11 | 897 | 153 | |
BC010887.1 | - | 10 - 11 | 1274 | 153 | |
BC019326.1 | - | 4 - 5 | 259 | 153 | |
CR457263.1 | - | 10 - 11 | 813 | 153 |