The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037808.2 | NP_001032897.1 | 7 - 8 | 827 | 77 |
NM_001199142.1 | NP_001186071.1 | 7 - 8 | 947 | 77 | |
NM_001267574.2 | NP_001254503.1 | 6 - 7 | 916 | 77 | |
NM_001286478.1 | NP_001273407.1 | 7 - 8 | 748 | 77 | |
NM_003752.4 | NP_003743.1 | 7 - 8 | 778 | 77 | |
XM_017023814.1 | XP_016879303.1 | 7 - 8 | 733 | 77 | |
GenBank mRNA | AK000739.1 | - | 6 - 7 | 808 | 77 |
AK055366.1 | - | 7 - 8 | 647 | 77 | |
AK292155.1 | - | 7 - 8 | 947 | 77 | |
AK301183.1 | - | 6 - 7 | 686 | 77 | |
AK303790.1 | - | 7 - 8 | 691 | 77 | |
AK304500.1 | - | 6 - 7 | 457 | 77 | |
AY762099.1 | - | 7 - 8 | 690 | 77 | |
BC001571.1 | - | 7 - 8 | 712 | 77 | |
BC071705.1 | - | 7 - 8 | 706 | 77 | |
BC107692.1 | - | 7 - 8 | 738 | 77 | |
BC157842.1 | - | 7 - 8 | 836 | 77 | |
BC157849.1 | - | 6 - 7 | 676 | 77 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099661.2 | NP_001093131.1 | 7 - 8 | 779 | 77 |
NM_001317856.1 | NP_001304785.1 | 7 - 8 | 824 | 77 | |
NM_001317857.1 | NP_001304786.1 | 7 - 8 | 775 | 77 | |
XM_017023620.1 | XP_016879109.1 | 7 - 8 | 731 | 77 |