Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr.4: 128809623 - 128875224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287437.1 NP_001274366.1 9 - 10 1748 74
NM_001287439.1 NP_001274368.1 9 - 10 1902 74
NM_001287440.1 NP_001274369.1 9 - 10 1699 74
NM_001287442.1 NP_001274371.1 9 - 10 1656 74
NM_001287443.1 NP_001274372.1 9 - 10 1660 74
NM_199320.3 NP_955352.1 9 - 10 1784 74
XM_005263232.1 XP_005263289.1 9 - 10 1626 74
GenBank mRNA AB058710.1 - 5 - 6 1084 74
AK127326.1 - 9 - 10 1556 74
AK304235.1 - 9 - 10 1617 74
AK304815.1 - 9 - 10 1573 74

View Full Product Details