The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145056.1 | NP_001138528.1 | 11 - 12 | 1033 | 94 |
NM_020428.3 | NP_065161.3 | 11 - 12 | 1092 | 94 | |
XM_005259997.1 | XP_005260054.1 | 11 - 12 | 1092 | 94 | |
XM_005259999.1 | XP_005260056.1 | 11 - 12 | 1033 | 94 | |
GenBank mRNA | AK027519.1 | - | 11 - 12 | 977 | 94 |
AK309801.1 | - | 9 - 10 | 862 | 94 | |
AK314582.1 | - | 11 - 12 | 1034 | 94 | |
AL832517.1 | - | 1 - 2 | 138 | 94 | |
AL832978.1 | - | 8 - 9 | 776 | 94 | |
BC010617.2 | - | 8 - 9 | 783 | 94 | |
BC040556.1 | - | 11 - 12 | 1034 | 94 | |
BC042692.1 | - | 11 - 12 | 1034 | 94 | |
FN668568.1 | - | 11 - 12 | 1092 | 94 |