The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000898.4 | NP_000889.3 | 12 - 13 | 1416 | 72 |
XM_005272608.3 | XP_005272665.1 | 14 - 15 | 1516 | 72 | |
XM_017029523.1 | XP_016885012.1 | 14 - 15 | 1566 | 72 | |
XM_017029524.1 | XP_016885013.1 | 13 - 14 | 1376 | 72 | |
GenBank mRNA | AK294306.1 | - | 13 - 14 | 1334 | 72 |
AK312679.1 | - | 12 - 13 | 1391 | 72 | |
BC022494.1 | - | 12 - 13 | 1362 | 72 | |
M69177.1 | - | 12 - 13 | 1317 | 72 | |
S62734.1 | - | 12 - 13 | 1240 | 72 |