The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271844.1 | NP_001258773.1 | 3 - 4 | 382 | 85 |
NM_001271845.1 | NP_001258774.1 | 3 - 4 | 421 | 85 | |
NM_001271846.1 | NP_001258775.1 | 3 - 4 | 421 | 85 | |
NM_019020.3 | NP_061893.2 | 7 - 8 | 1537 | 85 | |
XM_005257049.2 | XP_005257106.1 | 7 - 8 | 1548 | 85 | |
XM_005257050.3 | XP_005257107.1 | 7 - 8 | 1551 | 85 | |
XM_005257052.3 | XP_005257109.1 | 7 - 8 | 1554 | 85 | |
XM_006721694.3 | XP_006721757.1 | 7 - 8 | 1554 | 85 | |
XM_006721695.3 | XP_006721758.1 | 7 - 8 | 1554 | 85 | |
XM_011524322.2 | XP_011522624.1 | 7 - 8 | 1512 | 85 | |
XM_011524323.2 | XP_011522625.2 | 5 - 6 | 1462 | 85 | |
XM_011524324.2 | XP_011522626.1 | 3 - 4 | 1006 | 85 | |
XM_017024185.1 | XP_016879674.1 | 7 - 8 | 1551 | 85 | |
XM_017024186.1 | XP_016879675.1 | 5 - 6 | 1289 | 85 | |
XM_017024187.1 | XP_016879676.1 | 6 - 7 | 1529 | 85 | |
XM_017024188.1 | XP_016879677.1 | 5 - 6 | 669 | 85 | |
GenBank mRNA | AB449902.1 | - | 6 - 7 | 1411 | 85 |
BC001525.2 | - | 3 - 4 | 392 | 85 | |
BC028290.1 | - | 7 - 8 | 1527 | 85 | |
BC036947.1 | - | 3 - 4 | 426 | 85 | |
CR936670.1 | - | 3 - 4 | 387 | 85 |