Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr.17: 80169992 - 80209331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257970.1 NP_001244899.1 10 - 11 1663 102
NM_024110.4 NP_077015.2 10 - 11 1663 102
NM_052819.2 NP_438170.1 7 - 8 944 102
NR_047566.1 11 - 12 1660 102
XM_011525212.1 XP_011523514.1 12 - 13 2184 102
XM_011525213.1 XP_011523515.1 12 - 13 1926 102
XM_011525215.1 XP_011523517.1 11 - 12 1986 102
XM_011525216.1 XP_011523518.1 11 - 12 1686 102
XM_011525217.1 XP_011523519.1 11 - 12 1742 102
XM_011525218.2 XP_011523520.1 11 - 12 1725 102
XM_017025058.1 XP_016880547.1 12 - 13 2184 102
GenBank mRNA AF322642.1 - 10 - 11 1701 102
AK054723.1 - 2 - 3 379 102
AK091123.1 - 11 - 12 1618 102
AK296762.1 - 7 - 8 884 102
AK309299.1 - 6 - 7 974 102
AX746790.1 - 11 - 12 1618 102
AY032927.1 - 9 - 10 1498 102
BC001326.1 - 7 - 8 944 102
BC018142.1 - 10 - 11 1672 102
EU652409.1 - 9 - 10 1495 102

View Full Product Details