Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr.17: 80169992 - 80209331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024110.4 NP_077015.2 15 - 16 2391 95
NR_047566.1 16 - 17 2388 95
XM_011525212.1 XP_011523514.1 17 - 18 2912 95
XM_011525213.1 XP_011523515.1 17 - 18 2654 95
XM_011525215.1 XP_011523517.1 16 - 17 2714 95
XM_011525216.1 XP_011523518.1 16 - 17 2414 95
XM_011525217.1 XP_011523519.1 16 - 17 2470 95
XM_011525218.2 XP_011523520.1 16 - 17 2453 95
XM_017025057.1 XP_016880546.1 17 - 18 2909 95
GenBank mRNA AF322642.1 - 15 - 16 2429 95
AK091123.1 - 16 - 17 2346 95
AK296762.1 - 11 - 12 1485 95
AX746790.1 - 16 - 17 2346 95
AY032927.1 - 14 - 15 2226 95

View Full Product Details