Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr.17: 80169992 - 80209331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257970.1 NP_001244899.1 2 - 3 381 62
NM_024110.4 NP_077015.2 2 - 3 381 62
NR_047566.1 3 - 4 446 62
XM_011525212.1 XP_011523514.1 4 - 5 902 62
XM_011525213.1 XP_011523515.1 4 - 5 644 62
XM_011525215.1 XP_011523517.1 3 - 4 704 62
XM_011525216.1 XP_011523518.1 3 - 4 404 62
XM_011525217.1 XP_011523519.1 3 - 4 460 62
XM_011525218.2 XP_011523520.1 3 - 4 443 62
XM_017025057.1 XP_016880546.1 4 - 5 902 62
XM_017025058.1 XP_016880547.1 4 - 5 902 62
GenBank mRNA AF322642.1 - 2 - 3 419 62
AK091123.1 - 3 - 4 404 62
AX746790.1 - 3 - 4 404 62
AY032927.1 - 1 - 2 216 62
BC018142.1 - 2 - 3 390 62
EU652409.1 - 1 - 2 213 62

View Full Product Details