The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145252.1 | NP_001138724.1 | 1 - 2 | 203 | 105 |
NM_002621.2 | NP_002612.1 | 2 - 3 | 319 | 105 | |
GenBank mRNA | AK122955.1 | - | 1 - 2 | 128 | 105 |
AK310695.1 | - | 1 - 2 | 198 | 105 | |
BC015756.1 | - | 1 - 2 | 137 | 105 | |
DB146591.1 | - | 1 - 2 | 204 | 105 | |
KU178249.1 | - | 1 - 2 | 77 | 105 | |
KU178250.1 | - | 1 - 2 | 77 | 105 | |
X57748.1 | - | 1 - 2 | 142 | 105 |