The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175854.7 | NP_787050.6 | 15 - 16 | 2342 | 88 |
XM_005266332.3 | XP_005266389.1 | 15 - 16 | 2389 | 88 | |
XM_005266333.3 | XP_005266390.1 | 14 - 15 | 2028 | 88 | |
XM_005266334.1 | XP_005266391.1 | 15 - 16 | 1870 | 88 | |
XM_011535032.2 | XP_011533334.1 | 15 - 16 | 2336 | 88 | |
XM_011535034.2 | XP_011533336.1 | 15 - 16 | 2110 | 88 | |
XM_017020529.1 | XP_016876018.1 | 16 - 17 | 1882 | 88 | |
XM_017020531.1 | XP_016876020.1 | 12 - 13 | 1682 | 88 | |
XM_017020532.1 | XP_016876021.1 | 15 - 16 | 1840 | 88 | |
XM_017020533.1 | XP_016876022.1 | 13 - 14 | 1973 | 88 | |
GenBank mRNA | AB107584.1 | - | 13 - 14 | 1590 | 88 |
AB109552.1 | - | 14 - 15 | 1752 | 88 | |
AK131556.1 | - | 9 - 10 | 980 | 88 | |
BC024318.1 | - | 8 - 9 | 856 | 88 | |
BC128179.1 | - | 15 - 16 | 1870 | 88 | |
BC128180.1 | - | 13 - 14 | 1590 | 88 | |
BX647740.1 | - | 2 - 3 | 233 | 88 |