Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WWC1
Entrez Gene ID
23286
Gene Name
WW and C2 domain containing 1
Gene Aliases
HBEBP3, HBEBP36, KIBRA, MEMRYQTL, PPP1R168
Chromosome Location
Chr.5: 168292060 - 168472303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161661.1 NP_001155133.1 9 - 10 1277 65
NM_001161662.1 NP_001155134.1 9 - 10 1277 65
NM_015238.2 NP_056053.1 9 - 10 1277 65
XM_005265850.1 XP_005265907.1 9 - 10 1234 65
XM_005265853.2 XP_005265910.1 9 - 10 1234 65
XM_011534485.1 XP_011532787.1 11 - 12 1417 65
XM_011534486.1 XP_011532788.1 11 - 12 1417 65
XM_011534487.1 XP_011532789.1 11 - 12 1417 65
XM_011534488.1 XP_011532790.1 11 - 12 1417 65
XM_011534489.1 XP_011532791.1 11 - 12 1417 65
XM_011534490.1 XP_011532792.1 11 - 12 1417 65
XM_011534491.1 XP_011532793.1 11 - 12 1417 65
XM_017009276.1 XP_016864765.1 11 - 12 1417 65
XM_017009277.1 XP_016864766.1 11 - 12 1780 65
XM_017009278.1 XP_016864767.1 9 - 10 1669 65
GenBank mRNA AB020676.1 - 4 - 5 513 65
AF506799.1 - 9 - 10 1250 65
AF530058.1 - 4 - 5 581 65
AK126376.1 - 9 - 10 1250 65
AK296323.1 - 9 - 10 1187 65
BX640827.1 - 9 - 10 1081 65

View Full Product Details