Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
Chromosome Location
Chr.17: 19237377 - 19336715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148921.3 NP_683723.2 3 - 4 1046 55
GenBank mRNA AF062084.1 - 1 - 2 598 55
AK001996.1 - 1 - 2 765 55
AK024115.1 - 3 - 4 1026 55
BC070036.1 - 3 - 4 1011 55
BC093972.1 - 1 - 2 671 55
BC093974.1 - 1 - 2 671 55
BC143264.1 - 1 - 2 672 55

View Full Product Details