The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042517.1 | NP_001035982.1 | 16 - 17 | 2180 | 158 |
NM_001258366.1 | NP_001245295.1 | 15 - 16 | 2147 | 158 | |
NM_001258367.1 | NP_001245296.1 | 14 - 15 | 2042 | 158 | |
NM_001258368.1 | NP_001245297.1 | 14 - 15 | 1970 | 158 | |
NM_001258369.1 | NP_001245298.1 | 16 - 17 | 2180 | 158 | |
NM_001258370.1 | NP_001245299.1 | 10 - 11 | 1351 | 158 | |
NM_030932.3 | NP_112194.2 | 10 - 11 | 1351 | 158 | |
XM_006719876.1 | XP_006719939.1 | 10 - 11 | 1331 | 158 | |
XM_011535258.2 | XP_011533560.1 | 16 - 17 | 2123 | 158 | |
XM_011535263.2 | XP_011533565.1 | 16 - 17 | 2123 | 158 | |
XM_011535265.2 | XP_011533567.1 | 16 - 17 | 2123 | 158 | |
XM_017020789.1 | XP_016876278.1 | 16 - 17 | 2123 | 158 | |
GenBank mRNA | AB244756.1 | - | 15 - 16 | 1928 | 158 |
AB244757.1 | - | 14 - 15 | 1823 | 158 | |
AB244758.1 | - | 14 - 15 | 1751 | 158 | |
AK092024.1 | - | 17 - 18 | 2102 | 158 | |
AK298509.1 | - | 16 - 17 | 1966 | 158 | |
AK307652.1 | - | 10 - 11 | 1331 | 158 | |
AK316474.1 | - | 16 - 17 | 2093 | 158 | |
AL137718.1 | - | 10 - 11 | 1323 | 158 | |
AY750055.1 | - | 16 - 17 | 2053 | 158 | |
AY818645.1 | - | 16 - 17 | 1961 | 158 | |
BC034952.1 | - | 10 - 11 | 1355 | 158 | |
BC048963.1 | - | 10 - 11 | 1307 | 158 | |
BC068504.1 | - | 10 - 11 | 1307 | 158 |