The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042517.1 | NP_001035982.1 | 10 - 11 | 1338 | 128 |
NM_001258366.1 | NP_001245295.1 | 9 - 10 | 1305 | 128 | |
NM_001258367.1 | NP_001245296.1 | 8 - 9 | 1200 | 128 | |
NM_001258368.1 | NP_001245297.1 | 8 - 9 | 1128 | 128 | |
NM_001258369.1 | NP_001245298.1 | 10 - 11 | 1338 | 128 | |
NM_001258370.1 | NP_001245299.1 | 4 - 5 | 509 | 128 | |
NM_030932.3 | NP_112194.2 | 4 - 5 | 509 | 128 | |
XM_006719876.1 | XP_006719939.1 | 4 - 5 | 489 | 128 | |
XM_011535258.2 | XP_011533560.1 | 10 - 11 | 1281 | 128 | |
XM_011535263.2 | XP_011533565.1 | 10 - 11 | 1281 | 128 | |
XM_011535265.2 | XP_011533567.1 | 10 - 11 | 1281 | 128 | |
XM_017020789.1 | XP_016876278.1 | 10 - 11 | 1281 | 128 | |
GenBank mRNA | AB244756.1 | - | 9 - 10 | 1086 | 128 |
AB244757.1 | - | 8 - 9 | 981 | 128 | |
AB244758.1 | - | 8 - 9 | 909 | 128 | |
AK092024.1 | - | 11 - 12 | 1260 | 128 | |
AK298509.1 | - | 10 - 11 | 1124 | 128 | |
AK307652.1 | - | 4 - 5 | 489 | 128 | |
AK316474.1 | - | 10 - 11 | 1251 | 128 | |
AL137718.1 | - | 4 - 5 | 481 | 128 | |
AY750055.1 | - | 10 - 11 | 1211 | 128 | |
AY818645.1 | - | 10 - 11 | 1119 | 128 | |
BC034952.1 | - | 4 - 5 | 513 | 128 | |
BC048963.1 | - | 4 - 5 | 465 | 128 | |
BC068504.1 | - | 4 - 5 | 465 | 128 | |
BX649186.1 | - | 10 - 11 | 1280 | 128 |