Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PITHD1
Entrez Gene ID
57095
Gene Name
PITH domain containing 1
Gene Aliases
C1orf128, HT014, TXNL1CL
Chromosome Location
Chr.1: 23778386 - 23788232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020362.4 NP_065095.2 4 - 5 546 111
XM_011541804.1 XP_011540106.1 3 - 4 471 111
GenBank mRNA AF218024.1 - 4 - 5 527 111
AF221595.1 - 4 - 5 538 111
AF271784.1 - 4 - 5 542 111
AK296662.1 - 3 - 4 471 111
AK310330.1 - 4 - 5 493 111
AK313005.1 - 4 - 5 514 111
BC017208.2 - 4 - 5 507 111

View Full Product Details