Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 14 - 15 1909 76
XM_005273013.4 XP_005273070.1 13 - 14 1741 76
XM_005273018.1 XP_005273075.1 13 - 14 1748 76
XM_005273021.4 XP_005273078.1 14 - 15 1681 76
XM_005273022.3 XP_005273079.1 11 - 12 1530 76
XM_006711727.3 XP_006711790.1 14 - 15 1795 76
XM_006711728.3 XP_006711791.1 13 - 14 1666 76
XM_011544021.2 XP_011542323.1 15 - 16 1891 76
XM_011544022.2 XP_011542324.1 15 - 16 1966 76
XM_011544023.2 XP_011542325.1 15 - 16 1891 76
XM_011544024.2 XP_011542326.1 15 - 16 1891 76
XM_011544030.2 XP_011542332.1 7 - 8 2442 76
XM_017000103.1 XP_016855592.1 15 - 16 1891 76
XM_017000104.1 XP_016855593.1 13 - 14 1741 76
XM_017000106.1 XP_016855595.1 14 - 15 1877 76
XM_017000107.1 XP_016855596.1 15 - 16 1793 76
XM_017000108.1 XP_016855597.1 17 - 18 2340 76
GenBank mRNA AF250731.1 - 14 - 15 1740 76
AK297105.1 - 10 - 11 1328 76
AK299212.1 - 14 - 15 1858 76
AK299249.1 - 11 - 12 1501 76
BC045832.1 - 3 - 4 348 76

View Full Product Details