Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 1 - 2 234 82
XM_005273013.4 XP_005273070.1 1 - 2 195 82
XM_005273022.3 XP_005273079.1 1 - 2 238 82
XM_005273023.4 XP_005273080.1 1 - 2 911 82
XM_011544022.2 XP_011542324.1 1 - 2 195 82
XM_017000104.1 XP_016855593.1 1 - 2 195 82
XM_017000105.1 XP_016855594.1 1 - 2 221 82
XM_017000108.1 XP_016855597.1 1 - 2 195 82
GenBank mRNA AF161348.1 - 1 - 2 211 82
AF250731.1 - 1 - 2 65 82
AK299212.1 - 1 - 2 183 82
BC032454.1 - 1 - 2 177 82
DC326670.1 - 1 - 2 234 82

View Full Product Details