The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133369.2 | NP_588610.2 | 10 - 11 | 1825 | 101 |
XM_006714927.1 | XP_006714990.1 | 10 - 11 | 1805 | 101 | |
XM_006714928.1 | XP_006714991.1 | 11 - 12 | 1973 | 101 | |
XM_011534686.1 | XP_011532988.1 | 11 - 12 | 1973 | 101 | |
GenBank mRNA | BC009333.2 | - | 4 - 5 | 731 | 101 |
BC122556.1 | - | 4 - 5 | 702 | 101 | |
BC157824.1 | - | 10 - 11 | 1708 | 101 |