Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
Chromosome Location
Chr.5: 141430591 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003735.2 NP_003726.1 1 - 2 2596 131
GenBank mRNA AB011160.1 - 1 - 2 2569 131
AF152321.1 - 1 - 2 2427 131

View Full Product Details