Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM3
Entrez Gene ID
127294
Gene Name
myomesin 3
Gene Aliases
-
Chromosome Location
Chr.1: 24056041 - 24112175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152372.3 NP_689585.3 14 - 15 1782 79
GenBank mRNA AK125972.1 - 12 - 13 1824 79
AK128233.1 - 6 - 7 842 79
BC067101.1 - 14 - 15 1792 79
BX537911.1 - 13 - 14 1506 79
BX647453.1 - 14 - 15 1743 79
BX647455.1 - 14 - 15 1782 79
BX647644.1 - 14 - 15 1743 79

View Full Product Details