The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152372.3 | NP_689585.3 | 27 - 28 | 70 | |
GenBank mRNA | AK128233.1 | - | 19 - 20 | 70 | |
BC014671.1 | - | 1 - 2 | 44 | 70 | |
BC067101.1 | - | 27 - 28 | 70 | ||
BX537911.1 | - | 26 - 27 | 70 | ||
BX647453.1 | - | 27 - 28 | 70 | ||
BX647455.1 | - | 27 - 28 | 70 | ||
BX647644.1 | - | 27 - 28 | 70 |