Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
Chromosome Location
Chr.1: 20742677 - 20787809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016287.3 NP_057371.2 3 - 4 336 112
XM_005245875.4 XP_005245932.1 3 - 4 911 112
XM_005245876.3 XP_005245933.1 3 - 4 444 112
XM_005245877.4 XP_005245934.1 3 - 4 772 112
XM_005245878.4 XP_005245935.1 3 - 4 708 112
XM_005245879.4 XP_005245936.1 2 - 3 235 112
XM_011541532.1 XP_011539834.1 3 - 4 428 112
XM_011541533.1 XP_011539835.1 3 - 4 625 112
XM_011541534.2 XP_011539836.1 2 - 3 245 112
XM_017001393.1 XP_016856882.1 3 - 4 391 112
XM_017001394.1 XP_016856883.1 3 - 4 756 112
GenBank mRNA AF113534.1 - 2 - 3 266 112
AK023129.1 - 3 - 4 336 112
AK291252.1 - 3 - 4 314 112
AK304065.1 - 2 - 3 212 112
BC032139.2 - 3 - 4 423 112
BC046170.1 - 3 - 4 371 112

View Full Product Details