Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
Chromosome Location
Chr.1: 20742677 - 20787809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016287.3 NP_057371.2 4 - 5 481 121
XM_005245875.4 XP_005245932.1 4 - 5 1056 121
XM_005245876.3 XP_005245933.1 4 - 5 589 121
XM_005245877.4 XP_005245934.1 4 - 5 917 121
XM_005245878.4 XP_005245935.1 4 - 5 853 121
XM_005245879.4 XP_005245936.1 3 - 4 380 121
XM_011541532.1 XP_011539834.1 4 - 5 573 121
XM_011541533.1 XP_011539835.1 4 - 5 770 121
XM_011541534.2 XP_011539836.1 3 - 4 390 121
XM_017001393.1 XP_016856882.1 4 - 5 536 121
XM_017001394.1 XP_016856883.1 4 - 5 901 121
GenBank mRNA AF113534.1 - 3 - 4 411 121
AK023129.1 - 4 - 5 481 121
AK291252.1 - 4 - 5 459 121
AK304065.1 - 3 - 4 357 121
BC046170.1 - 4 - 5 516 121
CK000759.1 - 1 - 2 143 121

View Full Product Details