Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
Chromosome Location
Chr.1: 20742677 - 20787809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016287.3 NP_057371.2 10 - 11 1280 74
XM_005245875.4 XP_005245932.1 10 - 11 1855 74
XM_005245876.3 XP_005245933.1 10 - 11 1388 74
XM_005245877.4 XP_005245934.1 10 - 11 1716 74
XM_005245878.4 XP_005245935.1 10 - 11 1652 74
XM_005245879.4 XP_005245936.1 9 - 10 1179 74
XM_011541532.1 XP_011539834.1 10 - 11 1372 74
XM_011541533.1 XP_011539835.1 10 - 11 1569 74
XM_011541534.2 XP_011539836.1 9 - 10 1189 74
XM_017001393.1 XP_016856882.1 10 - 11 1335 74
XM_017001394.1 XP_016856883.1 10 - 11 1700 74
GenBank mRNA AF113534.1 - 9 - 10 1213 74
AK023129.1 - 10 - 11 1280 74
AK291252.1 - 10 - 11 1258 74
AK303456.1 - 3 - 4 396 74
AK304065.1 - 9 - 10 1156 74
AL833978.1 - 6 - 7 1096 74
BC045660.1 - 6 - 7 1912 74

View Full Product Details